Assay Details
Target Gene Details
Entrez Gene ID: | 54351 |
Gene Name: | elongator acetyltransferase complex subunit 5 |
Gene Aliases: |
Derp6, Rai12 |
Location: |
Chr.11:69968224-69981294 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Elp5 | NM_001253700.1 | NP_001240629.1 | ||
NM_018740.2 | NP_061210.2 | |||
AF004107.1 | AAC16252.1 | |||
AK011130.1 | ||||
AK013662.1 | BAB28944.1 | |||
AK152582.1 | BAE31332.1 | |||
BC003752.1 | AAH03752.1 | |||
BE862527.1 | ||||
CD741073.1 | ||||
CD776986.1 |