Assay Details
Target Gene Details
Entrez Gene ID: | 13650 |
Gene Name: | rhomboid family 1 (Drosophila) |
Gene Aliases: |
C16ORF8, Dist, Dist1, Egfr-rs, mKIAA4242 |
Location: |
Chr.11:32209584-32222331 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rhbdf1 | NM_001291818.1 | NP_001278747.1 | ||
NM_010117.2 | NP_034247.2 | |||
AK144437.1 | ||||
AK191461.1 | ||||
AK220566.1 | BAD90543.1 | |||
BC027346.1 | AAH27346.1 | |||
BY733688.1 | ||||
CD554689.1 | ||||
CN664154.1 |
More Information
Set Membership: |
Intragenic Non-exonic |