Assay Details
Target Gene Details
Entrez Gene ID: | 18103 |
Gene Name: | NME/NM23 nucleoside diphosphate kinase 2 |
Gene Aliases: |
NM23-H2, NM23B, nm23-M2 |
Location: |
Chr.11:93949814-93956256 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nme2 | NM_001077529.2 | NP_001070997.1 | ||
NM_008705.5 | NP_032731.1 | |||
AK012447.1 | BAB28246.1 | |||
AK150419.1 | BAE29543.1 | |||
AK151262.1 | BAE30250.1 | |||
AK169120.1 | BAE40900.1 | |||
AK210541.1 | ||||
AK216511.1 | ||||
BC066995.1 | AAH66995.1 | |||
BC086892.1 | AAH86892.1 | |||
BC086893.1 | AAH86893.1 | |||
BY514605.1 | ||||
EF437324.1 | ||||
X68193.1 | CAA48275.1 |
More Information
Set Membership: |
Intragenic Non-exonic |