Assay Details
Target Gene Details
Entrez Gene ID: | 109342 |
Gene Name: | solute carrier family 5 (sodium/glucose cotransporter), member 10 |
Gene Aliases: |
C330021F16Rik, SGLT5 |
Location: |
Chr.11:61672782-61720811 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Slc5a10 | NM_001033227.2 | 1 | 150 | NP_001028399.1 |
AK144033.1 | 1 | 151 | BAE25665.1 |
More Information
Set Membership: |
Intragenic Exonic |