Assay Details
Target Gene Details
Entrez Gene ID: | 18453 |
Gene Name: | prolyl 4-hydroxylase, beta polypeptide |
Gene Aliases: |
ERp59, PDI, Pdia1, Thbp |
Location: |
Chr.11:120560304-120572936 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
P4hb | NM_011032.2 | 8 | 1255 | NP_035162.1 |
AK141304.1 | 8 | 1192 | BAE24642.1 | |
AK146542.1 | 8 | 1193 | BAE27247.1 | |
AK149605.1 | 8 | 1194 | BAE28987.1 | |
AK149965.1 | 8 | 1193 | BAE29199.1 | |
AK150002.1 | 8 | 1219 | BAE29230.1 | |
AK150422.1 | 8 | 1193 | BAE29545.1 | |
AK150592.1 | 8 | 1193 | ||
AK150755.1 | 8 | 1179 | BAE29825.1 | |
AK150805.1 | 8 | 1193 | BAE29868.1 | |
AK151351.1 | 8 | 1187 | BAE30327.1 | |
AK151546.1 | 8 | 1193 | BAE30492.1 | |
AK152141.1 | 8 | 1193 | BAE30979.1 | |
AK152217.1 | 8 | 1193 | BAE31045.1 | |
AK152791.1 | 8 | 1193 | ||
AK152834.1 | 8 | 1193 | BAE31534.1 | |
AK153365.1 | 8 | 1193 | ||
AK159606.1 | 8 | 1193 | BAE35225.1 | |
AK159729.1 | 8 | 1193 | BAE35323.1 | |
AK159965.1 | 8 | 1193 | BAE35520.1 | |
AK161537.1 | BAE36449.1 | |||
AK167757.1 | BAE39791.1 | |||
AK167796.1 | 8 | 1193 | BAE39826.1 | |
AK168330.1 | 8 | 1191 | BAE40268.1 | |
AK168336.1 | 8 | 1175 | BAE40274.1 | |
AK168612.1 | 8 | 1193 | BAE40478.1 | |
AK168893.1 | 8 | 1193 | BAE40710.1 | |
AK169265.1 | 8 | 1190 | BAE41026.1 | |
AK170603.1 | 8 | 1190 | BAE41907.1 | |
BC008549.1 | 8 | 1181 | AAH08549.1 | |
BC093512.1 | 8 | 1172 | AAH93512.1 | |
J05185.1 | 8 | 1255 | AAA39906.1 | |
X06453.1 | 8 | 1195 | CAA29759.1 |
More Information
Set Membership: |
Intragenic Exonic |