Assay Details
Target Gene Details
Entrez Gene ID: | 170472 |
Gene Name: | RecQ protein-like 5 |
Gene Aliases: |
RecQ5, Recq5b, Recql5b |
Location: |
Chr.11:115892595-115933531 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Recql5 | NM_130454.2 | 2 | 767 | NP_569721.1 |
AB038773.1 | 2 | 101 | BAD11130.1 | |
AB059999.1 | 2 | 281 | BAB79232.1 | |
AK029060.1 | 2 | 763 | ||
AK083513.1 | 2 | 763 |
More Information
Set Membership: |
Intragenic Exonic |