Assay Details
Target Gene Details
Entrez Gene ID: | 170472 |
Gene Name: | RecQ protein-like 5 |
Gene Aliases: |
RecQ5, Recq5b, Recql5b |
Location: |
Chr.11:115892595-115933531 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Recql5 | NM_130454.2 | 1 | 506 | NP_569721.1 |
AB059999.1 | BAB79232.1 | |||
AK029060.1 | 1 | 502 | ||
AK083513.1 | 1 | 502 |
More Information
Set Membership: |
Intragenic Exonic |