Assay Details
Target Gene Details
Entrez Gene ID: | 68106 |
Gene Name: | 5'-nucleotidase, cytosolic IIIB |
Gene Aliases: |
2610037D24Rik, AI841843, C330027I04Rik, Nt5c3l |
Location: |
Chr.11:100422321-100441827 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nt5c3b | NM_001102650.1 | NP_001096120.1 | ||
NM_026561.4 | NP_080837.3 | |||
AK006972.1 | BAB24814.1 | |||
AK082816.1 | BAC38632.1 | |||
AK148222.1 | BAE28422.1 | |||
AK162770.1 | BAE37053.1 | |||
AK211190.1 | ||||
BC015307.1 | AAH15307.1 | |||
BY047327.1 |
More Information
Set Membership: |
Intragenic Non-exonic |