Assay Details
Target Gene Details
Entrez Gene ID: | 68106 |
Gene Name: | 5'-nucleotidase, cytosolic IIIB |
Gene Aliases: |
2610037D24Rik, AI841843, C330027I04Rik, Nt5c3l |
Location: |
Chr.11:100422321-100441827 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Nt5c3b | AK006972.1 | 2 | 146 | BAB24814.1 |
More Information
Set Membership: |
Intragenic Non-exonic |