Assay Details
Target Gene Details
Entrez Gene ID: | 19193 |
Gene Name: | pipecolic acid oxidase |
Gene Aliases: |
LPIPOX, Pso |
Location: |
Chr.11:77880614-77893872 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pipox | NM_008952.2 | NP_032978.2 | ||
AK008555.2 | BAB25741.1 | |||
AK135009.1 | 1 | 1574 | ||
AK149488.1 | BAE28913.1 | |||
BB561815.1 | ||||
BC013525.1 | AAH13525.1 | |||
BY708439.1 | ||||
U94700.1 | AAC39948.1 |
More Information
Set Membership: |
Intragenic Non-exonic |