Assay Details
Target Gene Details
Entrez Gene ID: | 19193 |
Gene Name: | pipecolic acid oxidase |
Gene Aliases: |
LPIPOX, Pso |
Location: |
Chr.11:77880614-77893872 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pipox | NM_008952.2 | 1 | 106 | NP_032978.2 |
AK008555.2 | 1 | 67 | BAB25741.1 | |
AK135009.1 | ||||
AK149488.1 | 1 | 55 | BAE28913.1 | |
BB561815.1 | 1 | 106 | ||
BC013525.1 | AAH13525.1 | |||
BY708439.1 | 1 | 67 | ||
U94700.1 | AAC39948.1 |
More Information
Set Membership: |
Intragenic Exonic |