Assay Details
Target Gene Details
Entrez Gene ID: | 52898 |
Gene Name: | ribonuclease, RNase K |
Gene Aliases: |
2310033H11Rik, AA571397, D11Bwg0434e |
Location: |
Chr.11:70238123-70239852 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rnasek | NM_173742.3 | NP_776103.1 | ||
AK003548.1 | ||||
AK009592.1 | BAC25261.1 | |||
AK018709.1 | ||||
AK171975.1 | BAE42755.1 | |||
BC021602.1 | AAH21602.2 | |||
BC021603.1 | AAH21603.2 | |||
BC024417.1 | AAH24417.1 | |||
CJ181610.1 |