Assay Details
Target Gene Details
Entrez Gene ID: | 57376 |
Gene Name: | SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily e, member 1 |
Gene Aliases: |
2810417B20Rik, 5830412H02Rik, 9030408N19Rik, Baf27 |
Location: |
Chr.11:99209047-99231339 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Smarce1 | NM_020618.4 | NP_065643.1 | ||
AK018493.1 | ||||
AK076177.1 | BAC36233.1 | |||
BC047141.1 | AAH47141.1 | |||
BC061498.1 | AAH61498.1 | |||
BC065043.1 | AAH65043.1 |
More Information
Set Membership: |
Intragenic Non-exonic |