Assay Details
Target Gene Details
Entrez Gene ID: | 209011 |
Gene Name: | sirtuin 7 |
Gene Aliases: |
- |
Location: |
Chr.11:120618372-120625002 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sirt7 | NM_153056.2 | NP_694696.2 | ||
AB221687.1 | ||||
AK135484.1 | ||||
AY251540.1 | AAP83960.1 | |||
BC026403.1 | AAH26403.1 | |||
BC026650.1 | AAH26650.1 | |||
BC040759.1 |