Assay Details
Target Gene Details
Entrez Gene ID: | 216850 |
Gene Name: | KDM1 lysine (K)-specific demethylase 6B |
Gene Aliases: |
1700064E03Rik, BC038313, Jmjd3, mKIAA0346 |
Location: |
Chr.11:69398508-69418148 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kdm6b | NM_001017426.1 | 2 | 208 | NP_001017426.1 |
BC075632.1 | 3 | 384 | AAH75632.1 |
More Information
Set Membership: |
Intragenic Exonic |