Assay Details
Target Gene Details
Entrez Gene ID: | 16515 |
Gene Name: | potassium inwardly-rectifying channel, subfamily J, member 12 |
Gene Aliases: |
IRK-2, IRK2, Kir2.2, MB-IRK2 |
Location: |
Chr.11:61022564-61073269 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kcnj12 | NM_001267593.1 | NP_001254522.1 | ||
NM_010603.6 | 1 | 126 | NP_034733.4 | |
AK017299.1 | 1 | 127 | BAB30679.1 | |
AK032271.1 | BAC27788.1 |
More Information
Set Membership: |
Intragenic Exonic |