Assay Details
Target Gene Details
Entrez Gene ID: | 84653 |
Gene Name: | hairy and enhancer of split 7 (Drosophila) |
Gene Aliases: |
bHLHb37 |
Location: |
Chr.11:69118711-69124054 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hes7 | NM_033041.4 | NP_149030.2 | ||
AB049065.1 | BAB39526.1 | |||
AK050791.1 | BAC34412.1 | |||
BC134378.1 |
More Information
Set Membership: |
Intragenic Non-exonic |