Assay Details
Target Gene Details
Entrez Gene ID: | 14799 |
Gene Name: | glutamate receptor, ionotropic, AMPA1 (alpha 1) |
Gene Aliases: |
2900051M01Rik, Glr-1, Glr1, GluA1, GluR-A, GluRA, Glur-1, Glur1, HIPA1, gluR-K1 |
Location: |
Chr.11:57011571-57330244 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gria1 | NM_001113325.2 | NP_001106796.1 | ||
NM_001252403.1 | NP_001239332.1 | |||
NM_008165.4 | NP_032191.2 | |||
AB102773.1 | ||||
AB102774.1 | ||||
AK049958.1 | ||||
AK135107.1 | ||||
BC056397.1 | AAH56397.1 | |||
BC060702.1 | AAH60702.1 | |||
X57497.1 | CAA40734.1 |
More Information
Set Membership: |
Intragenic Non-exonic |