Assay Details
Target Gene Details
Entrez Gene ID: | 14534 |
Gene Name: | K(lysine) acetyltransferase 2A |
Gene Aliases: |
1110051E14Rik, AW212720, Gcn5, Gcn5l2, mmGCN5 |
Location: |
Chr.11:100704746-100712467 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Kat2a | NM_001038010.2 | NP_001033099.1 | ||
NM_020004.5 | NP_064388.2 | |||
AF254441.1 | AAF70497.1 | |||
AK137358.1 | BAE23321.1 | |||
AK145205.1 | BAE26296.1 | |||
AK147042.1 | ||||
AK150225.1 | BAE29392.1 | |||
AK154866.1 | ||||
AK155347.1 | BAE33207.1 | |||
AK155791.1 | ||||
AK155842.1 | BAE33458.1 | |||
AK158079.1 | BAE34351.1 | |||
AK185926.1 | ||||
BC063752.1 | AAH63752.1 |