Assay Details
Target Gene Details
Entrez Gene ID: | 17913 |
Gene Name: | myosin IC |
Gene Aliases: |
C80397, MMIb, MYO1E, NMI, mm1beta, myr2 |
Location: |
Chr.11:75650493-75674643 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 13 - Intron 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Myo1c | NM_001080774.1 | NP_001074243.1 | ||
NM_001080775.1 | NP_001074244.1 | |||
NM_008659.3 | NP_032685.1 | |||
AK004743.1 | BAB23524.1 | |||
AK154294.1 | BAE32495.1 | |||
AK155530.1 | BAE33311.1 | |||
AK165680.1 | ||||
AK166638.1 | ||||
AK171107.1 | BAE42253.1 | |||
AY007255.1 | AAG02570.1 | |||
BC021481.1 | AAH21481.1 | |||
U96723.1 | AAC53264.1 | |||
X99638.1 | CAA67956.1 |
More Information
Set Membership: |
Intragenic Non-exonic |