Assay Details
Target Gene Details
Entrez Gene ID: | 78304 |
Gene Name: | N(alpha)-acetyltransferase 38, NatC auxiliary subunit |
Gene Aliases: |
1500034E06Rik, Lsmd1 |
Location: |
Chr.11:69395791-69396671 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Naa38 | NM_030083.2 | NP_084359.1 | ||
AK018782.1 | BAB31406.1 | |||
BC048499.1 | ||||
BC062267.1 | AAH62267.1 |
More Information
Set Membership: |
Intragenic Non-exonic |