Assay Details
Target Gene Details
Entrez Gene ID: | 74302 |
Gene Name: | myotubularin related protein 3 |
Gene Aliases: |
1700092A20Rik, AI255150, AW557713, FYVE-DSP1, ZFYVE10, mKIAA0371 |
Location: |
Chr.11:4480868-4594815 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mtmr3 | NM_028860.2 | NP_083136.2 | ||
AK141138.1 | ||||
BC032166.1 | AAH32166.1 | |||
BC081544.1 | AAH81544.1 | |||
CJ048010.1 |
More Information
Set Membership: |
Intragenic Non-exonic |