Assay Details
Target Gene Details
Entrez Gene ID: | 268395 |
Gene Name: | N-methylpurine-DNA glycosylase |
Gene Aliases: |
9830006D05, AI326268, APNG, Aag, Mid1 |
Location: |
Chr.11:32226425-32232702 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mpg | NM_010822.3 | 1 | 154 | NP_034952.2 |
AK018694.1 | 1 | 134 | ||
AK076201.1 | 1 | 149 | ||
AK088878.1 | 1 | 109 | ||
AK134900.1 | 1 | 121 | ||
AK155766.1 | 1 | 155 | ||
AK156067.1 | 1 | 121 | BAE33569.1 | |
BC014754.1 | 1 | 148 | AAH14754.1 | |
BY309585.1 | 1 | 154 | ||
U10420.1 | 1 | 147 | AAA19487.1 |
More Information
Set Membership: |
Intragenic Exonic |