Assay Details
Target Gene Details
Entrez Gene ID: | 16399 |
Gene Name: | integrin alpha 2b |
Gene Aliases: |
AI172977, CD41, CD41B, GpIIb, alphaIIb |
Location: |
Chr.11:102453297-102469883 on Build GRCm38 |
Assay Gene Location: | Within Exon 29 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Itga2b | NM_010575.2 | 29 | 3164 | NP_034705.2 |
AF045019.1 | 6 | 628 | AAD02339.1 | |
AF170316.1 | 29 | 3025 | AAD56216.1 | |
AK140749.1 | 24 | 2816 | ||
AK142289.1 | 29 | 3026 | BAE25013.1 | |
AK154619.1 | 29 | 3165 | BAE32718.1 | |
AK156872.1 | 28 | 3345 | ||
AK157012.1 | 28 | 3349 | ||
AK158443.1 | 29 | 3016 | ||
AK161727.1 | 7 | 1258 | ||
BC120493.1 | 29 | 3014 | ||
BC137570.1 | 29 | 3014 |
More Information
Set Membership: |
Intragenic Exonic |