Assay Details
Target Gene Details
Entrez Gene ID: | 16399 |
Gene Name: | integrin alpha 2b |
Gene Aliases: |
AI172977, CD41, CD41B, GpIIb, alphaIIb |
Location: |
Chr.11:102453297-102469883 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Itga2b | NM_010575.2 | NP_034705.2 | ||
AF170316.1 | AAD56216.1 | |||
AK048700.1 | BAC33425.1 | |||
AK140749.1 | ||||
AK142289.1 | BAE25013.1 | |||
AK154619.1 | BAE32718.1 | |||
AK156872.1 | ||||
AK157012.1 | ||||
AK158443.1 | ||||
BC120493.1 | ||||
BC137570.1 |
More Information
Set Membership: |
Intragenic Non-exonic |