Assay Details
Target Gene Details
Entrez Gene ID: | 14783 |
Gene Name: | growth factor receptor bound protein 10 |
Gene Aliases: |
5730571D09Rik, AI325020, Meg1, mKIAA0207 |
Location: |
Chr.11:11930499-12037422 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Grb10 | NM_001177629.1 | NP_001171100.1 | ||
NM_010345.4 | NP_034475.2 | |||
AF022072.1 | AAB72103.1 | |||
AK030727.1 | BAC27100.1 | |||
AK032927.1 | BAC28088.1 | |||
AK163841.1 | BAE37514.1 | |||
AK169000.1 | BAE40799.1 | |||
BC016111.1 | AAH16111.1 | |||
BC053842.1 | AAH53842.1 |
More Information
Set Membership: |
Intragenic Non-exonic |