Assay Details
Target Gene Details
Entrez Gene ID: | 20185 |
Gene Name: | nuclear receptor co-repressor 1 |
Gene Aliases: |
5730405M06Rik, A230020K14Rik, N-CoR, RIP13, Rxrip13, mKIAA1047 |
Location: |
Chr.11:62315643-62457332 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 42 - Exon 43 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ncor1 | NM_001252313.1 | NP_001239242.1 | ||
NM_011308.3 | NP_035438.3 | |||
AB093281.1 | BAC41465.1 | |||
AK034988.1 | 1 | 2169 | ||
AK038502.1 | 1 | 118 | ||
AK052753.1 | 1 | 492 | ||
AK137632.1 | BAE23439.1 | |||
BC086657.1 | AAH86657.1 | |||
L78294.1 | AAL40135.1 | |||
U22016.1 | AAC52168.1 | |||
U35312.1 | AAB17125.1 |
More Information
Set Membership: |
Intragenic Non-exonic |