Assay Details
Target Gene Details
Entrez Gene ID: | 20185 |
Gene Name: | nuclear receptor co-repressor 1 |
Gene Aliases: |
5730405M06Rik, A230020K14Rik, N-CoR, RIP13, Rxrip13, mKIAA1047 |
Location: |
Chr.11:62315643-62457332 on Build GRCm38 |
Assay Gene Location: | Within Exon 37 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ncor1 | NM_001252313.1 | 35 | 5433 | NP_001239242.1 |
NM_011308.3 | 35 | 5232 | NP_035438.3 | |
AB093281.1 | 17 | 3173 | BAC41465.1 | |
AK137632.1 | 9 | 1551 | BAE23439.1 | |
BC086657.1 | 35 | 5198 | AAH86657.1 | |
L78294.1 | 14 | 2240 | AAL40135.1 | |
U22016.1 | AAC52168.1 | |||
U35312.1 | 34 | 5476 | AAB17125.1 |
More Information
Set Membership: |
Intragenic Exonic |