Assay Details
Target Gene Details
Entrez Gene ID: | 17882 |
Gene Name: | myosin, heavy polypeptide 2, skeletal muscle, adult |
Gene Aliases: |
MHC2A, MyHC-IIa, Myh2a, Myhs-f, Myhs-f1, Myhsf1 |
Location: |
Chr.11:67171027-67197517 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Myh2 | NM_001039545.2 | NP_001034634.2 |