Assay Details
Target Gene Details
Entrez Gene ID: | 16428 |
Gene Name: | IL2 inducible T cell kinase |
Gene Aliases: |
Emt, Tcsk, Tsk |
Location: |
Chr.11:46325148-46389515 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Itk | NM_001281965.1 | 13 | 1508 | NP_001268894.1 |
NM_001281966.1 | 13 | 1499 | NP_001268895.1 | |
NM_001281968.1 | 13 | 1487 | NP_001268897.1 | |
NM_010583.3 | 13 | 1490 | NP_034713.2 | |
AK083380.1 | 13 | 1462 | ||
AK153613.1 | 13 | 1446 | BAE32118.1 | |
BC128374.1 | 13 | 1416 | ||
BC128375.1 | 13 | 1413 | ||
D14042.1 | 13 | 1430 | BAA03129.1 | |
L00619.1 | 13 | 1495 | AAA39337.1 | |
L05631.1 | 13 | 1456 | AAA40518.1 | |
L10628.1 | 13 | 1458 |