Assay Details
Target Gene Details
Entrez Gene ID: | 15384 |
Gene Name: | heterogeneous nuclear ribonucleoprotein A/B |
Gene Aliases: |
3010025C11Rik, CBF-A, Cgbfa, Hnrpab |
Location: |
Chr.11:51600100-51606881 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hnrnpab | NM_001048061.1 | NP_001041526.1 | ||
NM_010448.3 | NP_034578.1 | |||
AK013384.1 | BAB28821.1 | |||
AK076132.1 | BAC36208.1 | |||
AK165609.1 | BAE38291.1 | |||
BC043069.1 | AAH43069.1 | |||
CJ166869.1 | ||||
D90151.1 | BAA14181.1 |