Assay Details
Target Gene Details
Entrez Gene ID: | 171285 |
Gene Name: | hepatitis A virus cellular receptor 2 |
Gene Aliases: |
TIM-3, Tim3, Timd3 |
Location: |
Chr.11:46454547-46481255 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Havcr2 | NM_134250.2 | NP_599011.2 | ||
AF450241.1 | AAL65156.1 | |||
AK148474.1 | BAE28574.1 | |||
AY553334.1 | AAS59531.1 |
More Information
Set Membership: |
Intragenic Non-exonic |