Assay Details
Target Gene Details
Entrez Gene ID: | 17879 |
Gene Name: | myosin, heavy polypeptide 1, skeletal muscle, adult |
Gene Aliases: |
A530084A17Rik, IId, IId/x, MHC-2X/D, MHC2X/D, MYHC-IIX, MdMs, MyHC-IId/x, MyHC-IIx/d, Myhs-f, Myhs-f2, Myhsf2 |
Location: |
Chr.11:67200114-67224575 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 17 - Intron 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Myh1 | NM_030679.1 | NP_109604.1 | ||
AK041122.1 | BAC30831.1 | |||
BC108329.1 | AAI08330.1 | |||
BC150739.1 |