Assay Details
Target Gene Details
Entrez Gene ID: | 66720 |
Gene Name: | kelch-like 10 |
Gene Aliases: |
4921517C11Rik |
Location: |
Chr.11:100441924-100457027 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Klhl10 | NM_025727.3 | NP_080003.1 | ||
AK006288.1 | BAB24507.1 | |||
AK014906.1 | BAB29614.1 | |||
AK016593.1 | ||||
AK077202.1 | BAC36680.1 | |||
AY495337.1 | AAS91790.1 | |||
BC139267.1 | ||||
BC139268.1 |
More Information
Set Membership: |
Intragenic Non-exonic |