Assay Details
Target Gene Details
Entrez Gene ID: | 192897 |
Gene Name: | integrin beta 4 |
Gene Aliases: |
AA407042, C230078O20, CD104 |
Location: |
Chr.11:115974725-116008412 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Itgb4 | NM_001005608.2 | 20 | 2749 | NP_001005608.2 |
NM_133663.2 | 20 | 2749 | NP_598424.2 | |
AK048888.1 | 20 | 2633 | ||
AK147424.1 | 20 | 2749 | ||
BC059192.1 | 16 | 1944 | AAH59192.1 | |
L04678.1 | 21 | 2593 |
More Information
Set Membership: |
Intragenic Exonic |