Assay Details
Target Gene Details
Entrez Gene ID: | 192897 |
Gene Name: | integrin beta 4 |
Gene Aliases: |
AA407042, C230078O20, CD104 |
Location: |
Chr.11:115974725-116008412 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 14 - Intron 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Itgb4 | NM_001005608.2 | NP_001005608.2 | ||
NM_133663.2 | NP_598424.2 | |||
AK048888.1 | ||||
AK147424.1 | ||||
BC059192.1 | AAH59192.1 | |||
L04678.1 |
More Information
Set Membership: |
Intragenic Non-exonic |