Assay Details
Target Gene Details
Entrez Gene ID: | 268510 |
Gene Name: | mannoside acetylglucosaminyltransferase 5, isoenzyme B |
Gene Aliases: |
C330018B01, GnT-IX, mGnTVB |
Location: |
Chr.11:116918856-116986944 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mgat5b | NM_172948.3 | NP_766536.2 | ||
AB116028.1 | BAD08454.1 | |||
AB119127.1 | BAC87707.1 | |||
BC094604.1 | AAH94604.1 |
More Information
Set Membership: |
Intragenic Non-exonic |