Assay Details
Target Gene Details
Entrez Gene ID: | 103534 |
Gene Name: | mannoside acetylglucosaminyltransferase 4, isoenzyme B |
Gene Aliases: |
AA407995, GnTIVb |
Location: |
Chr.11:50225335-50235103 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Mgat4b | NM_145926.3 | 11 | 1593 | NP_666038.3 |
AB053218.1 | 11 | 1295 | BAC55019.1 | |
AK155336.1 | 11 | 1594 | BAE33202.1 | |
BC026638.1 | 6 | 701 | AAH26638.1 | |
BC031613.1 | 5 | 571 | AAH31613.1 | |
CX206051.1 | 3 | 352 |
More Information
Set Membership: |
Intragenic Exonic |