Assay Details
Target Gene Details
Entrez Gene ID: | 15239 |
Gene Name: | HGF-regulated tyrosine kinase substrate |
Gene Aliases: |
Hgr, Hrs, ZFYVE8 |
Location: |
Chr.11:120467635-120483984 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hgs | NM_001159328.1 | NP_001152800.1 | ||
NM_008244.3 | NP_032270.3 | |||
AK046299.1 | BAC32676.1 | |||
AK145031.1 | BAE26195.1 | |||
AK166441.1 | BAE38778.1 | |||
AK180016.1 | ||||
AK217735.1 | ||||
BC003239.1 | AAH03239.1 | |||
D50050.1 | BAA08768.1 |