Assay Details
Target Gene Details
Entrez Gene ID: | 67487 |
Gene Name: | DEAH (Asp-Glu-Ala-His) box polypeptide 40 |
Gene Aliases: |
2410016C14Rik, ARG147, DDX40, PAD |
Location: |
Chr.11:86768849-86807660 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dhx40 | NM_026191.2 | NP_080467.3 | ||
AK004610.1 | BAC25091.1 | |||
AK010512.1 | BAB26995.2 | |||
AK029935.1 | ||||
AK031000.1 | BAC27210.1 | |||
AK075991.1 | BAC36102.1 | |||
AK135592.1 | ||||
AK208938.1 | ||||
BC058276.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic |