Assay Details
Target Gene Details
Entrez Gene ID: | 574403 |
Gene Name: | family with sequence similarity 196, member B |
Gene Aliases: |
EG574403, Gm6041 |
Location: |
Chr.11:34314822-34422642 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fam196b | NM_001025382.2 | NP_001020553.1 | ||
BC072605.1 | AAH72605.1 |