Assay Details
Target Gene Details
Entrez Gene ID: | 13861 |
Gene Name: | eosinophil peroxidase |
Gene Aliases: |
EPO |
Location: |
Chr.11:87863997-87875536 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Epx | NM_007946.2 | NP_031972.2 | ||
BB843302.1 | ||||
D78353.1 | BAA11370.1 | |||
L77979.1 | AAB40403.1 |
More Information
Set Membership: |
Intragenic Non-exonic |