Assay Details
Target Gene Details
Entrez Gene ID: | 78394 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 52 |
Gene Aliases: |
2700029C06Rik, ROK1 |
Location: |
Chr.11:83942090-83963088 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ddx52 | NM_030096.2 | 8 | 1123 | NP_084372.2 |
AK080767.1 | 8 | 1061 | BAC38014.1 | |
AK133545.1 | 8 | 1094 | BAE21716.1 | |
AK141200.1 | 8 | 1097 | ||
AK166789.1 | 8 | 1049 | BAE39019.1 | |
AK195121.1 | ||||
BC029094.1 | 8 | 1035 | AAH29094.1 |