Assay Details
Target Gene Details
Entrez Gene ID: | 216797 |
Gene Name: | protease, serine 38 |
Gene Aliases: |
Gm249, Mpn2 |
Location: |
Chr.11:59372669-59376253 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Prss38 | NM_001045521.1 | 2 | 241 | NP_001038986.1 |
AK145810.1 | 2 | 246 | BAE26664.1 | |
BC147400.1 | 2 | 237 | ||
BC147401.1 | 2 | 237 | ||
BY736706.1 | 2 | 241 |
More Information
Set Membership: |
Intragenic Exonic |