Assay Details
Target Gene Details
Entrez Gene ID: | 327951 |
Gene Name: | cytochrome b5 domain containing 1 |
Gene Aliases: |
Gm740 |
Location: |
Chr.11:69390930-69395346 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cyb5d1 | NM_001045525.1 | NP_001038990.1 | ||
BC099556.1 | AAH99556.1 |