Assay Details
Target Gene Details
Entrez Gene ID: | 13385 |
Gene Name: | discs, large homolog 4 (Drosophila) |
Gene Aliases: |
Dlgh4, PSD-95, PSD95, SAP90, SAP90A |
Location: |
Chr.11:70018455-70045532 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dlg4 | NM_001109752.1 | NP_001103222.1 | ||
NM_007864.3 | NP_031890.1 | |||
BC014807.1 | AAH14807.1 | |||
BQ959940.1 | ||||
D50621.1 | BAA09297.1 |
More Information
Set Membership: |
Intragenic Non-exonic |