Assay Details
Target Gene Details
Entrez Gene ID: | 12464 |
Gene Name: | chaperonin containing Tcp1, subunit 4 (delta) |
Gene Aliases: |
2610204B21Rik, A45, C78323, Cctd |
Location: |
Chr.11:22990593-23003336 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cct4 | NM_009837.1 | NP_033967.1 | ||
AK010634.1 | BAB27078.1 | |||
AK146242.1 | BAE27006.1 | |||
AK150601.1 | BAE29693.1 | |||
AK152173.1 | BAE31005.1 | |||
AK152665.1 | BAE31400.1 | |||
AK163369.1 | ||||
AK165229.1 | BAE38089.1 | |||
AK167026.1 | BAE39199.1 | |||
AK167262.1 | BAE39376.1 | |||
AK167657.1 | ||||
AK167846.1 | BAE39866.1 | |||
AK169339.1 | BAE41091.1 | |||
AK200879.1 | ||||
AK206188.1 | ||||
AK208716.1 | ||||
AK215884.1 | ||||
BC054773.1 | AAH54773.1 | |||
L25913.1 | AAA37418.1 | |||
Z31554.1 | CAA83429.1 |
More Information
Set Membership: |
Intragenic Non-exonic |