Assay Details
Target Gene Details
Entrez Gene ID: | 216616 |
Gene Name: | epidermal growth factor-containing fibulin-like extracellular matrix protein 1 |
Gene Aliases: |
- |
Location: |
Chr.11:28853205-28926743 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Efemp1 | NM_146015.2 | 6 | 826 | NP_666127.2 |
AK077302.1 | 6 | 828 | BAC36738.1 | |
AY185605.1 | 5 | 651 | AAO37642.1 | |
AY251056.1 | 6 | 821 | AAP79577.1 | |
BC031184.1 | 3 | 447 | AAH31184.1 |
More Information
Set Membership: |
Intragenic Exonic |