Assay Details
Target Gene Details
Entrez Gene ID: | 13169 |
Gene Name: | drebrin-like |
Gene Aliases: |
Abp1, SH3P7, mAbp1 |
Location: |
Chr.11:5788483-5800980 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Dbnl | NM_001146308.1 | 4 | 366 | NP_001139780.1 |
NM_001146309.1 | 4 | 366 | NP_001139781.1 | |
NM_013810.3 | 4 | 366 | NP_038838.1 | |
AK046073.1 | 4 | 346 | BAC32592.1 | |
AK053796.1 | 4 | 362 | BAC35528.1 | |
AK078082.1 | 4 | 333 | BAC37118.1 | |
AK134136.1 | 4 | 333 | BAE22028.1 | |
AK134487.1 | 4 | 333 | BAE22160.1 | |
AK140752.1 | 4 | 333 | BAE24466.1 | |
AK142818.1 | 4 | 316 | BAE25201.1 | |
AK146920.1 | 4 | 333 | BAE27532.1 | |
AK151096.1 | 4 | 346 | BAE30108.1 | |
AK152110.1 | 4 | 331 | ||
AK152204.1 | 4 | 346 | BAE31033.1 | |
AK153072.1 | 4 | 346 | ||
AK153389.1 | 4 | 320 | BAE31953.1 | |
AK159984.1 | 4 | 343 | BAE35535.1 | |
AK161973.1 | 4 | 346 | ||
AK167956.1 | 4 | 306 | ||
AK168898.1 | 4 | 343 | BAE40714.1 | |
AK172471.1 | 4 | 323 | BAE43024.1 | |
AK199448.1 | ||||
AK203166.1 | ||||
AY098595.1 | 4 | 289 | AAM28340.1 | |
BC046430.1 | 4 | 331 | AAH46430.1 | |
BY293844.1 | ||||
U58884.1 | 4 | 319 | AAC52640.1 |
More Information
Set Membership: |
Intragenic Exonic |