Assay Details
Target Gene Details
Entrez Gene ID: | 216848 |
Gene Name: | chromodomain helicase DNA binding protein 3 |
Gene Aliases: |
2600010P09Rik, AF020312, Chd7, Prp7, Prp9-1 |
Location: |
Chr.11:69343273-69369426 on Build GRCm38 |
Assay Gene Location: | Within Exon 31 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Chd3 | NM_146019.4 | 31 | 4918 | NP_666131.3 |
BM950040.1 | ||||
BU702731.1 | 6 | 645 | ||
CB195281.1 | 2 | 201 | ||
CK389202.1 |